siRNA Details
| Gene Symbol: Magea8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020020.4 | NP_064404.1 | 4 | 400 | ||
| GenBank mRNA | AK145480.1 | - | 4 | 401 | ||
| BC119510.2 | - | 1 | 91 | 40055160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magea8| Gene Symbol: Magea8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020020.4 | NP_064404.1 | 4 | 400 | ||
| GenBank mRNA | AK145480.1 | - | 4 | 401 | ||
| BC119510.2 | - | 1 | 91 | 40055160 | ||