siRNA Details
| Gene Symbol: Dppa5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025274.3 | NP_079550.2 | 1 | 168 | ||
| GenBank mRNA | AF490349.1 | - | 1 | 124 | ||
| AK010519.1 | - | 1 | 125 | |||
| AK010584.1 | - | 1 | 124 | |||
| AK145761.1 | - | 1 | 127 | |||
| AV457404.1 | - | 1 | 170 | |||
| BC092353.1 | - | 1 | 113 | 30730511 | ||
| BC092354.1 | - | 1 | 121 | 30732471 | ||
| BC095979.1 | - | 1 | 113 | 30732937 | ||
| X57708.1 | - | 1 | 126 | |||
| Gene Symbol: Gm6083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG071895.2 | - | 1 | 272 | ||

Dppa5a
Genomic Map