siRNA Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025728.4 | NP_080004.1 | 2 | 227 | ||
| NM_029160.3 | NP_083436.2 | 11 | 1068 | |||
| GenBank mRNA | AF490391.1 | - | 11 | 1050 | ||
| AK014865.1 | - | 2 | 222 | |||
| AK015884.1 | - | 11 | 1069 | |||
| AK016352.1 | - | 2 | 195 | |||
| AK016618.1 | - | 2 | 214 | |||
| BC120668.1 | - | 11 | 1067 | 40129591 | ||
| BC145006.1 | - | 11 | 1067 | 9053554 | ||
| CJ046986.1 | - | 2 | 228 | |||

Spag16
Genomic Map