siRNA Details
Gene Symbol: Speer4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281511.1 | NP_001268440.1 | 5 | 1010 | ||
GenBank mRNA | BB015819.1 | - | 1 | 68 |
Gene Symbol: 4930572O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076955.1 | - | 5 | 982 |
Gene Symbol: Speer4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025759.3 | NP_080035.2 | 5 | 975 | ||
GenBank mRNA | AK017002.1 | - | 5 | 976 |
