siRNA Details
| Gene Symbol: Speer4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025759.3 | NP_080035.2 | 5 | 889 | ||
| GenBank mRNA | AK017002.1 | - | 5 | 890 | ||
| Gene Symbol: Speer4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281511.1 | NP_001268440.1 | 5 | 924 | ||

Speer4d
Genomic Map