siRNA Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026445.2 | NP_080721.1 | 9 | 1353 | ||
| GenBank mRNA | AK076022.1 | - | 9 | 1359 | ||
| AK138300.1 | - | 9 | 1344 | |||
| AK140169.1 | - | 10 | 1368 | |||
| AK157784.1 | - | 9 | 1354 | |||
| AK166216.1 | - | 9 | 1313 | |||

Sumf2
Genomic Map