siRNA Details
| Gene Symbol: Dpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026767.4 | NP_081043.1 | 2 | 439 | ||
| GenBank mRNA | AI527477.1 | - | 1 | 383 | ||
| AK003223.1 | - | 2 | 349 | |||
| BC037761.1 | - | 1 | 383 | |||
| CJ237809.1 | - | 2 | 439 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Dpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026767.4 | NP_081043.1 | 2 | 439 | ||
| GenBank mRNA | AI527477.1 | - | 1 | 383 | ||
| AK003223.1 | - | 2 | 349 | |||
| BC037761.1 | - | 1 | 383 | |||
| CJ237809.1 | - | 2 | 439 | |||
Dpm3
Genomic Map