siRNA Details
| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026914.1 | NP_081190.1 | 3 | 396 | ||
| GenBank mRNA | AK005345.1 | - | 3 | 388 | ||
| AK151325.1 | - | 3 | 364 | |||
| AK152795.1 | - | 3 | 364 | |||
| AK170899.1 | - | 3 | 386 | |||
| AK184297.1 | - | 2 | 180 | |||
| BC028457.1 | - | 3 | 396 | 1245697 | ||
| BC092006.1 | - | 3 | 351 | |||

Smdt1
Genomic Map