siRNA Details
Gene Symbol: Ptms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026988.2 | NP_081264.1 | 1, 2 | 398 | ||
GenBank mRNA | AK011360.1 | - | 1, 2 | 398 | ||
BC052699.1 | - | 1, 2 | 383 | 5715702 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026988.2 | NP_081264.1 | 1, 2 | 398 | ||
GenBank mRNA | AK011360.1 | - | 1, 2 | 398 | ||
BC052699.1 | - | 1, 2 | 383 | 5715702 |