siRNA Details
| Gene Symbol: Ikbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026166.2 | NP_080442.2 | 2 | 322 | ||
| NM_027078.2 | NP_081354.1 | 2 | 322 | |||
| GenBank mRNA | AK004670.1 | - | 2 | 280 | ||
| AK006283.1 | - | 2 | 290 | |||
| BC034890.1 | - | 2 | 284 | 1447362 | ||
| BC055834.1 | - | 2 | 273 | 3492302 | ||
| BY277133.1 | - | 2 | 354 | |||
| CF171082.1 | - | 2 | 304 | |||

Ikbip
Genomic Map