siRNA Details
| Gene Symbol: Rnaseh2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027187.3 | NP_081463.1 | 3 | 266 | ||
| GenBank mRNA | AK010292.1 | - | 3 | 256 | ||
| AK146518.1 | - | 3 | 267 | |||
| BC029252.1 | - | 3 | 296 | 3666709 | ||
| BC038158.1 | - | 3 | 340 | 1493985 | ||

Rnaseh2a
Genomic Map