siRNA Details
| Gene Symbol: Obox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145708.2 | NP_663754.2 | 4 | 1204 | ||
| GenBank mRNA | BC094912.1 | - | 4 | 1084 | ||
| DV647958.1 | - | 1 | 592 | |||
| Gene Symbol: Obox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027802.2 | NP_082078.1 | 4 | 1112 | ||
| GenBank mRNA | AK018362.1 | - | 4 | 1109 | ||
| AK136061.1 | - | 4 | 1102 | |||
| Gene Symbol: Obox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145709.2 | NP_663755.2 | 4 | 1180 | ||
| GenBank mRNA | BB701159.1 | - | 1 | 349 | ||
| BC052664.1 | - | 4 | 1184 | 30053785 | ||
| DV653414.1 | - | 2 | 745 | |||
| Gene Symbol: Obox7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038676.1 | NP_001033765.1 | 4 | 1148 | ||
| GenBank mRNA | BC099488.1 | - | 4 | 1148 | 30939375 | |

Obox2
Genomic Map