siRNA Details
| Gene Symbol: 1700001F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027940.2 | NP_082216.1 | 5 | 578 | ||
| GenBank mRNA | AK005587.1 | - | 5 | 579 | ||
| BC061060.1 | - | 5 | 560 | 6770724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001F09Rik| Gene Symbol: 1700001F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027940.2 | NP_082216.1 | 5 | 578 | ||
| GenBank mRNA | AK005587.1 | - | 5 | 579 | ||
| BC061060.1 | - | 5 | 560 | 6770724 | ||