siRNA Details
| Gene Symbol: Ccdc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028312.3 | NP_082588.2 | 6 | 437 | ||
| GenBank mRNA | AK012610.1 | - | 6 | 438 | ||
| BC026668.1 | - | 6 | 387 | 5006213 | ||
| BE627582.1 | - | 2 | 172 | |||
| CA450868.1 | - | 4 | 171 | |||

Ccdc12
Genomic Map