siRNA Details
| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029232.2 | NP_083508.1 | 3 | 207 | ||
| GenBank mRNA | AK016214.1 | - | 3 | 198 | ||
| BC050803.1 | - | 3 | 207 | 6771315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco5b| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029232.2 | NP_083508.1 | 3 | 207 | ||
| GenBank mRNA | AK016214.1 | - | 3 | 198 | ||
| BC050803.1 | - | 3 | 207 | 6771315 | ||