siRNA Details
| Gene Symbol: Slc35f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029238.2 | NP_083514.1 | 5 | 1049 | ||
| GenBank mRNA | AB041538.1 | - | 6 | 977 | ||
| AK016089.1 | - | 2 | 244 | |||
| AK034308.1 | - | 5 | 1051 | |||
| BC138981.1 | - | 5 | 948 | 8862003 | ||
| BC145720.1 | - | 5 | 948 | 40131023 | ||
 Back To Top
 Back To Top
 Slc35f4
Slc35f4 Genomic Map
Genomic Map Back To Top
 Back To Top