siRNA Details
| Gene Symbol: Speer4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029376.2 | NP_083652.2 | 4 | 605 | ||
| GenBank mRNA | AK006439.1 | - | 4 | 609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer4a| Gene Symbol: Speer4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029376.2 | NP_083652.2 | 4 | 605 | ||
| GenBank mRNA | AK006439.1 | - | 4 | 609 | ||