siRNA Details
| Gene Symbol: 1700010D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005817.1 | - | 1 | 90 | ||
| BC059715.1 | - | 1 | 93 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700010D01Rik| Gene Symbol: 1700010D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005817.1 | - | 1 | 90 | ||
| BC059715.1 | - | 1 | 93 | |||