siRNA Details
| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020026.1 | - | 5 | 634 | ||
| AK035837.1 | - | 1898 | ||||
| AK046610.1 | - | 5 | 662 | |||
| AK052819.1 | - | 1871 | ||||
| AK089083.1 | - | 5 | 475 | |||
| AK168294.1 | - | 6 | 922 | |||
| BC031464.1 | - | 5 | 619 | 4017482 | ||

Trappc6b
Genomic Map