siRNA Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030075.2 | NP_084351.1 | 7 | 1510 | ||
| GenBank mRNA | AK019849.1 | - | 7 | 1508 | ||
| AK154428.1 | - | 7 | 1611 | |||
| BC048918.1 | - | 2 | 473 | 6309434 | ||
| BC060089.1 | - | 2 | 814 | |||

Klhdc8b
Genomic Map