siRNA Details
| Gene Symbol: Hist3h2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018765.1 | - | 1 | 461 | ||
| BC051921.1 | - | 1 | 451 | 5684450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist3h2ba| Gene Symbol: Hist3h2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018765.1 | - | 1 | 461 | ||
| BC051921.1 | - | 1 | 451 | 5684450 | ||