siRNA Details
| Gene Symbol: 8030411F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030135.2 | NP_084411.1 | 1 | 119 | ||
| GenBank mRNA | AF440735.1 | - | 1 | 119 | ||
| AK020193.1 | - | 1 | 35 | |||
| BC132218.1 | - | 1 | 64 | 40130495 | ||
| BC132222.1 | - | 1 | 64 | 40130499 | ||
| BC145532.1 | - | 1 | 64 | |||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Rat: AF442205.
					            	This siRNA also targets the following transcripts in Rat: AF442205.Gene Ontology Categories:
 Back To Top
 Back To Top
 8030411F24Rik
8030411F24Rik Genomic Map
Genomic Map Back To Top
 Back To Top