siRNA Details
Gene Symbol: 8030411F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030135.2 | NP_084411.1 | 2 | 402 | ||
GenBank mRNA | AF440735.1 | - | 2 | 402 | ||
AK020193.1 | - | 2 | 318 | |||
BC132218.1 | - | 2 | 347 | 40130495 | ||
BC132222.1 | - | 2 | 347 | 40130499 |
