siRNA Details
| Gene Symbol: Psg28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054063.4 | NP_473404.3 | 5 | 1813 | ||
| GenBank mRNA | AF113598.1 | - | 5 | 1663 | ||
| BC099402.1 | - | 5 | 1799 | 30787049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg28| Gene Symbol: Psg28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054063.4 | NP_473404.3 | 5 | 1813 | ||
| GenBank mRNA | AF113598.1 | - | 5 | 1663 | ||
| BC099402.1 | - | 5 | 1799 | 30787049 | ||