siRNA Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138593.2 | NP_613059.2 | 10 | 1482 | ||
| GenBank mRNA | BC029178.1 | - | 2 | 155 | ||
| BC132266.1 | - | 2 | 153 | |||
| BC132268.1 | - | 2 | 153 | |||
| BM946683.1 | - | 4 | 350 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138593.2 | NP_613059.2 | 10 | 1482 | ||
| GenBank mRNA | BC029178.1 | - | 2 | 155 | ||
| BC132266.1 | - | 2 | 153 | |||
| BC132268.1 | - | 2 | 153 | |||
| BM946683.1 | - | 4 | 350 | |||
Larp7
Genomic Map