siRNA Details
| Gene Symbol: Olfr960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146279.1 | NP_666391.1 | 1 | 333 | ||
| GenBank mRNA | BC132468.1 | - | 1 | 353 | 40130745 | |
| BC132470.1 | - | 1 | 353 | 40130747 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr960| Gene Symbol: Olfr960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146279.1 | NP_666391.1 | 1 | 333 | ||
| GenBank mRNA | BC132468.1 | - | 1 | 353 | 40130745 | |
| BC132470.1 | - | 1 | 353 | 40130747 | ||