siRNA Details
| Gene Symbol: Olfr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146289.1 | NP_666401.1 | 1 | 447 | ||
| GenBank mRNA | BC132178.1 | - | 1 | 474 | 40130455 | |
| BC132180.1 | - | 1 | 474 | 40130457 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Olfr113
Olfr113| Gene Symbol: Olfr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146289.1 | NP_666401.1 | 1 | 447 | ||
| GenBank mRNA | BC132178.1 | - | 1 | 474 | 40130455 | |
| BC132180.1 | - | 1 | 474 | 40130457 | ||
 Back To Top
 Back To Top