siRNA Details
| Gene Symbol: Olfr110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146328.2 | NP_666440.2 | 2 | 962 | ||
| GenBank mRNA | BC132347.1 | - | 1 | 906 | 40130624 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr110| Gene Symbol: Olfr110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146328.2 | NP_666440.2 | 2 | 962 | ||
| GenBank mRNA | BC132347.1 | - | 1 | 906 | 40130624 | |