siRNA Details
Gene Symbol: Olfr1295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146403.1 | NP_666515.1 | 1 | 80 | ||
GenBank mRNA | BC125371.1 | - | 1 | 149 | 40129886 | |
BC131912.1 | - | 1 | 149 | 40130189 |
