siRNA Details
| Gene Symbol: Olfr1295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146403.1 | NP_666515.1 | 1 | 336 | ||
| GenBank mRNA | BC125371.1 | - | 1 | 405 | 40129886 | |
| BC131912.1 | - | 1 | 405 | 40130189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1295| Gene Symbol: Olfr1295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146403.1 | NP_666515.1 | 1 | 336 | ||
| GenBank mRNA | BC125371.1 | - | 1 | 405 | 40129886 | |
| BC131912.1 | - | 1 | 405 | 40130189 | ||