siRNA Details
Gene Symbol: Olfr462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146411.2 | NP_666523.2 | 1 | 409 | ||
GenBank mRNA | BC108939.1 | - | 409 | 40041186 | ||
BC108940.1 | - | 410 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146411.2 | NP_666523.2 | 1 | 409 | ||
GenBank mRNA | BC108939.1 | - | 409 | 40041186 | ||
BC108940.1 | - | 410 |