siRNA Details
| Gene Symbol: Olfr877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146417.1 | NP_666529.1 | 1 | 818 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Olfr877
Olfr877| Gene Symbol: Olfr877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146417.1 | NP_666529.1 | 1 | 818 | ||
 Back To Top
 Back To Top