siRNA Details
| Gene Symbol: WDFY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020830.4 | NP_065881.1 | 3 | 372 | ||
| GenBank mRNA | AB037856.1 | - | 3 | 285 | ||
| AF411977.1 | - | 3 | 269 | |||
| AJ310568.1 | - | 3 | 359 | |||
| AK022888.1 | - | 3 | 298 | |||
| AK023415.1 | - | 2 | 166 | |||
| BC040525.1 | - | 3 | 284 | 5285567 | ||
| BC065934.1 | - | 3 | 316 | 6164567 | ||
| DA775670.1 | - | 3 | 372 | |||

WDFY1
Genomic Map