siRNA Details
Gene Symbol: Olfr742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146430.2 | NP_666641.2 | 1 | 939 | ||
GenBank mRNA | BC132583.1 | - | 1 | 939 | 40130860 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
