siRNA Details
| Gene Symbol: Olfr1512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146432.2 | NP_666643.1 | 1 | 743 | ||
| GenBank mRNA | BC133656.1 | - | 1 | 743 | 40042187 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1512| Gene Symbol: Olfr1512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146432.2 | NP_666643.1 | 1 | 743 | ||
| GenBank mRNA | BC133656.1 | - | 1 | 743 | 40042187 | |