siRNA Details
| Gene Symbol: Olfr1231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146454.2 | NP_666665.2 | 1 | 876 | ||
| GenBank mRNA | BC104282.1 | - | 1 | 872 | ||
| BC104283.1 | - | 1 | 875 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1231| Gene Symbol: Olfr1231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146454.2 | NP_666665.2 | 1 | 876 | ||
| GenBank mRNA | BC104282.1 | - | 1 | 872 | ||
| BC104283.1 | - | 1 | 875 | |||