siRNA Details
Gene Symbol: Olfr1388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146467.1 | NP_666678.1 | 1 | 16 | ||
GenBank mRNA | BC104275.1 | - | 1 | 16 | 40041223 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146467.1 | NP_666678.1 | 1 | 16 | ||
GenBank mRNA | BC104275.1 | - | 1 | 16 | 40041223 |