siRNA Details
| Gene Symbol: Olfr1388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146467.1 | NP_666678.1 | 1 | 74 | ||
| GenBank mRNA | BC104274.1 | - | 1 | 73 | ||
| BC104275.1 | - | 1 | 74 | 40041223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1388| Gene Symbol: Olfr1388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146467.1 | NP_666678.1 | 1 | 74 | ||
| GenBank mRNA | BC104274.1 | - | 1 | 73 | ||
| BC104275.1 | - | 1 | 74 | 40041223 | ||