siRNA Details
| Gene Symbol: Olfr1393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146471.1 | NP_666682.1 | 1 | 308 | ||
| GenBank mRNA | BC104066.1 | - | 1 | 308 | 40041234 | |
| BC104067.1 | - | 1 | 308 | 40041236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1393| Gene Symbol: Olfr1393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146471.1 | NP_666682.1 | 1 | 308 | ||
| GenBank mRNA | BC104066.1 | - | 1 | 308 | 40041234 | |
| BC104067.1 | - | 1 | 308 | 40041236 | ||