siRNA Details
| Gene Symbol: Olfr90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146477.2 | NP_666688.2 | 1 | 615 | ||
| GenBank mRNA | BC119191.1 | - | 1 | 615 | 8733940 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr90| Gene Symbol: Olfr90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146477.2 | NP_666688.2 | 1 | 615 | ||
| GenBank mRNA | BC119191.1 | - | 1 | 615 | 8733940 | |