siRNA Details
Gene Symbol: Olfr183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146485.2 | NP_666696.2 | 2 | 347 | ||
GenBank mRNA | DR065568.1 | - | 2 | 366 | ||
DR065627.1 | - | 2 | 168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146485.2 | NP_666696.2 | 2 | 347 | ||
GenBank mRNA | DR065568.1 | - | 2 | 366 | ||
DR065627.1 | - | 2 | 168 |