siRNA Details
| Gene Symbol: Olfr1411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146490.1 | NP_666701.1 | 1 | 16 | ||
| GenBank mRNA | BC128013.1 | - | 1 | 20 | 40136041 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1411| Gene Symbol: Olfr1411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146490.1 | NP_666701.1 | 1 | 16 | ||
| GenBank mRNA | BC128013.1 | - | 1 | 20 | 40136041 | |