siRNA Details
| Gene Symbol: Olfr98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146510.1 | NP_666721.1 | 1 | 833 | ||
| GenBank mRNA | BC119060.1 | - | 1 | 920 | 8733809 | |
| BC120744.1 | - | 1 | 920 | 40129667 | ||
| Gene Symbol: Olfr97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC134363.1 | - | 840 | 40134689 | ||

Olfr98
Genomic Map