siRNA Details
| Gene Symbol: Olfr1170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146532.1 | NP_666743.1 | 1 | 554 | ||
| GenBank mRNA | BC127994.1 | - | 1 | 558 | 40135743 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1170| Gene Symbol: Olfr1170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146532.1 | NP_666743.1 | 1 | 554 | ||
| GenBank mRNA | BC127994.1 | - | 1 | 558 | 40135743 | |