siRNA Details
Gene Symbol: Olfr373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146539.2 | NP_666750.2 | 1 | 818 | ||
GenBank mRNA | BC106979.2 | - | 1 | 816 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146539.2 | NP_666750.2 | 1 | 818 | ||
GenBank mRNA | BC106979.2 | - | 1 | 816 |