siRNA Details
Gene Symbol: Olfr270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146607.1 | NP_666818.1 | 1 | 900 | ||
GenBank mRNA | BC127971.1 | - | 1 | 904 | 40135114 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146607.1 | NP_666818.1 | 1 | 900 | ||
GenBank mRNA | BC127971.1 | - | 1 | 904 | 40135114 |