siRNA Details
| Gene Symbol: Olfr357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146623.1 | NP_666834.1 | 1 | 333 | ||
| GenBank mRNA | BC119361.1 | - | 1 | 418 | 8734110 | |
| BC119363.1 | - | 1 | 418 | 8734112 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr357| Gene Symbol: Olfr357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146623.1 | NP_666834.1 | 1 | 333 | ||
| GenBank mRNA | BC119361.1 | - | 1 | 418 | 8734110 | |
| BC119363.1 | - | 1 | 418 | 8734112 | ||