siRNA Details
| Gene Symbol: Olfr350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146627.1 | NP_666838.1 | 1 | 406 | ||
| GenBank mRNA | BC106790.1 | - | 1 | 406 | ||
| BC106791.1 | - | 1 | 406 | |||
| BC119406.2 | - | 1 | 406 | 40041609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr350| Gene Symbol: Olfr350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146627.1 | NP_666838.1 | 1 | 406 | ||
| GenBank mRNA | BC106790.1 | - | 1 | 406 | ||
| BC106791.1 | - | 1 | 406 | |||
| BC119406.2 | - | 1 | 406 | 40041609 | ||