siRNA Details
| Gene Symbol: LRCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243963.1 | NP_001230892.1 | 14 | 1697 | ||
| NM_020871.3 | NP_065922.3 | 14 | 1697 | |||
| XM_006724724.3 | XP_006724787.2 | 14 | 1679 | |||
| XM_017029696.1 | XP_016885185.1 | 14 | 1706 | |||
| XM_017029697.1 | XP_016885186.1 | 14 | 1707 | |||
| GenBank mRNA | AB040928.1 | - | 10 | 878 | ||
| AK022128.1 | - | 6 | 453 | |||
| BC125223.1 | - | 2 | 104 | |||
| BC125224.1 | - | 14 | 1690 | 40124466 | ||
| BX647193.1 | - | 14 | 1697 | |||

LRCH2
Genomic Map