siRNA Details
| Gene Symbol: Olfr237-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146654.1 | NP_666865.1 | 1 | 332 | ||
| GenBank mRNA | BC120515.1 | - | 1 | 377 | 40129438 | |
| BC120519.1 | - | 1 | 377 | 40129442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr237-ps1| Gene Symbol: Olfr237-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146654.1 | NP_666865.1 | 1 | 332 | ||
| GenBank mRNA | BC120515.1 | - | 1 | 377 | 40129438 | |
| BC120519.1 | - | 1 | 377 | 40129442 | ||